missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-L-threonine 98.0+%, TCI America™

Numéro de catalogue. B33985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B33985G 5 g
B33981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B33985G Fournisseur TCI America Code fournisseur B33985G
Il en reste null

Identifiants chimiques

CAS 4378-13-6
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00037766
InChI Key NMJINEMBBQVPGY-RITPCOANSA-N
Synonym h-thr tbu-oh, o-tert-butyl-l-threonine, 2s,3r-2-amino-3-tert-butoxy butanoic acid, l-threonine, o-1,1-dimethylethyl, o-t-butyl-l-threonine, 2s,3r-2-amino-3-2-methylpropan-2-yl oxy butanoic acid, o-tert.-butylthreonin, o-tert.butyl-l-threonine, h-ser trt-2-chlorotrityl resin, h-thr tbu-2-chlorotrityl resin
PubChem CID 6951357
IUPAC Name (2S,3R)-2-amino-3-(tert-butoxy)butanoic acid
SMILES C[C@@H](OC(C)(C)C)[C@H](N)C(O)=O

Spécifications

Melting Point 260°C
Color White-Yellow
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-tert-Butyl-L-threonine