missing translation for 'onlineSavingsMsg'
Learn More

o-Terphenyl 99.0+%, TCI America™

Numéro de catalogue. T001925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T001925G 25 g
T0019100G 100 g
T0019500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. T001925G Fournisseur TCI America Code fournisseur T001925G
Il en reste null

Identifiants chimiques

CAS 84-15-1
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00003055
InChI Key OIAQMFOKAXHPNH-UHFFFAOYSA-N
Synonym o-terphenyl, 1,1':2',1-terphenyl, terphenyl, chlorinated, 1,1'-biphenyl, 2-phenyl, caswell no. 672b, o-diphenylbenzene, polychlorinated terphenyl, polychlorinated terphenyls, unii-w5675r7kvw, ccris 1655
PubChem CID 6766
IUPAC Name 1,2-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2C3=CC=CC=C3

Spécifications

Melting Point 58°C
Color White
Boiling Point 332°C
Quantity 25 g
UN Number 3077
Formula Weight 230.31
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material o-Terphenyl
EINECSNumber (4)-0017
RTECSNumber WZ6472000
TSCA Yes