missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ O'RangeRuler DNA Ladders, ready-to-use
GREENER_CHOICE

Numéro de catalogue. p-200113683
Change view
Click to view available options
Size Range:
10 to 100 bp
10 to 150 bp
100 to 500 bp
100 to 6000 bp
20 to 300 bp
200 to 3000 bp
50 to 1000 bp
500 to 6000 bp
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size Range
FERSM1303 10 to 100 bp
FERSM1313 10 to 150 bp
FERSM1323 20 to 300 bp
FERSM0613 50 to 1000 bp
FERSM0623 100 to 500 bp
FERSM0653 100 to 6000 bp
FERSM0633 200 to 3000 bp
FERSM0643 500 to 6000 bp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Numéro de catalogue. FERSM1303 Fournisseur Thermo Scientific™ Code fournisseur SM1303
Il en reste null

Thermo Scientific O'RangeRuler DNA Ladders is a step ladder designed for precise sizing of DNA fragments in agarose or non-denaturing polyacrylamide gels.

Thermo Scientific O'RangeRuler DNA Ladders are designed for precise sizing of PCR products and other double-stranded DNA fragments in agarose or non-denaturing polyacrylamide gels.

O'RangeRuler series is characterized by its 'step ladder' design. The chromatography-purified DNA fragments are precisely separated by a consistent increment (e.g., 5 bp, 10 bp, 20 bp, 50 bp, 100 bp, 200 bp, or 500 bp). The design enables accurate and precise sizing of DNA fragments. Not recommended for DNA quantification.

All O'RangeRuler DNA Ladders are supplied with 6X Orange DNA Loading Dye for sample DNA. Orange G dye can be used to monitor DNA migration in agarose or polyacrylamide gels and migrates at 50 bp in a 1% agarose gel.

Highlights

  • Step ladders available in a wide range of increments from 5-to-500 bp
  • Evenly spaced reference bands
  • Sharp bands, easy-to-remember fragment sizes
  • Ready-to-use—premixed with 6X Orange Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use O'RangeRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

Spécifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Orange G
Ready to Load Yes
Size Range 10 to 100 bp
Gel Compatibility Agarose Gels
Quantity 50 μg
Product Line O'GeneRuler, O’RangeRuler
Product Type DNA Ladder

For Research Use Only. Not for use in diagnostic procedures.