missing translation for 'onlineSavingsMsg'
Learn More

o-Phenanthroline , 0.1% Aqueous, for Iron, Certified, LabChem™

Catalog No. LC181501
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC181501 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC181501 Supplier LabChem Supplier No. LC181501
Only null left

Chemical Identifiers

CAS 7732-18-5 , 5144-89-8 , 7647-01-0
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Color Colorless
CAS Max % 99.8, 0.1, 0.1
Linear Formula C12H8N2·H2O
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 198.22
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Suitability Passes Test
Decomposition Information Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material o-Phenanthroline, 0.1% Aqueous, for Iron
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature