missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Phénanthroline, alcoolique à 0,1%, certifié, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | o-Phenanthroline |
| Nom Remarque | 0,1% alcoolique |
| CAS % maximal | 0.1, 89.91, 4.995, 4.995 |
| Renseignements sur la décomposition | Monoxyde de carbone; Le dioxyde de carbone; Peut libérer des gaz inflammables |
| Couleur | Incolore |
| Emballage | Bouteille en polyéthylène |
| Formule linéaire | C12H8N2·H2O |
| Quantité | 500 mL |
| Numéro UN | UN1170 |
| Renseignements sur la solubilité | Soluble dans l’eau |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Harmful if swallowed. May cause cancer. Suspected of damaging fertility or the unborn child. Causes damage to organs (central nervous system, optic nerve) (oral). |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Keep away from heat, sparks, open flames, hot surfaces. No smoking. Keep container tightly closed. Ground/bond container and receiving equipment. Use explosion-proof electrical, ventilating, lighting equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Do not breathe mist, vapors, spray. Do not eat, drink or smoke when using this product. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. If exposed or concerned: Get medical advice/attention. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish. Store locked up in cool, well-ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Température ambiante |