missing translation for 'onlineSavingsMsg'
Learn More

O-Ethylhydroxylamine hydrochloride, 99.0-101.0 wt. % (AT), MilliporeSigma™ Supelco™

Catalog No. 111013947
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013947 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013947 Supplier Merck Emd Millipore Supplier No. 4350410G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00012956
Synonym Ethoxyamine hydrochloride

Specifications

Melting Point 130°C to 133°C
CAS 3332-29-4
Linear Formula CH3 CH2 ONH2 · HCl
Quantity 10 g
Formula Weight 97.54
Percent Purity 99.0 to 101.0 wt. % (AT)
Grade For GC derivatization