missing translation for 'onlineSavingsMsg'
Learn More

o-Chloranil 97,0+%, TCI America™

Numéro de catalogue. T09705G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T09705G 5 g
T097025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T09705G Fournisseur TCI America Code fournisseur T09705G

Identifiants chimiques

CAS 2435-53-2
Molecular Formula C6Cl4O2
Molecular Weight (g/mol) 245.86
MDL Number MFCD00001646
InChI Key VRGCYEIGVVTZCC-UHFFFAOYSA-N
Synonym o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil
PubChem CID 73252
IUPAC Name tétrachlorocyclohexa-3,5-diène-1,2-dione
SMILES ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl

Spécifications

Melting Point 128°C
Quantity 5 g
Formula Weight 245.86
Percent Purity ≥97.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material o-Chloranil