missing translation for 'onlineSavingsMsg'
Learn More

O-Benzylhydroxylamine hydrochloride, 99%

Catalog No. p-7023636
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1739306 5 g
AAA1739314 25 g
AAA1739322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1739306 Supplier Thermo Scientific Chemicals Supplier No. A1739306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2687-43-6 | C7H10ClNO | 159.613 g/mol

For use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For use in the formation of nitrones from carbonyl compounds and their use in cycloaddition reactions. Used as a reagent used in the synthesis of hydroxylamines and hydroxyamates. Effective reagent used to prepare α-hydroxybenzylamines from α-hydroxyketones.

Solubility
Soluble in DMSO and methanol.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.

Chemical Identifiers

CAS 2687-43-6
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00012952
InChI Key HYDZPXNVHXJHBG-UHFFFAOYSA-N
Synonym o-benzylhydroxylamine hydrochloride, benzyloxyamine hydrochloride, o-benzylhydroxylamine hcl, o-benzyl-hydroxylamine hydrochloride, hydroxylamine, o-phenylmethyl-, hydrochloride, hydroxylamine, o-benzyl-, hydrochloride, o-benzyl-hydroxylamine hcl, o-benzyl hydroxylamine hydrochloride, aminooxy methyl benzene hydrochloride, oxybenzylamine, chloride
PubChem CID 102312
IUPAC Name O-benzylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)CON.Cl

Specifications

Melting Point ∼238°C (sublimation)
Quantity 5 g
Beilstein 3595440
Sensitivity Hygroscopic
Solubility Information Soluble in DMSO and methanol.
Formula Weight 159.62
Percent Purity 99%
Chemical Name or Material O-Benzylhydroxylamine hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-249-0
RTECSNumber NC3040000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only