missing translation for 'onlineSavingsMsg'
Learn More

O-Acetylsalicylic acid, 99%

CAS: 50-78-2 | C9H8O4 | 180.16 g/mol

Supplier:  Thermo Scientific Chemicals A124880E

 View more versions of this product

Catalog No. AAA124880E


Only null left
Explore available promotions

Description

Description

O-Acetylsalicylic acid is the prototypical analgesic used in the treatment of mild to moderate pain. acts as an inhibitor of cyclooxygenase which results in the inhibition of the biosynthesis of prostaglandins. Aspirin also inhibits platelet aggregation and is used in the prevention of arterial and venous thrombosis. Along with sodium salicylate (sc-3520), aspirin has been shown to block the long terminal repeat from the human immunodeficiency virus and the Ig κ enhancer from NFκB dependent transcription.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Acetylsalicylic acid is the prototypical analgesic used in the treatment of mild to moderate pain. acts as an inhibitor of cyclooxygenase which results in the inhibition of the biosynthesis of prostaglandins. Aspirin also inhibits platelet aggregation and is used in the prevention of arterial and venous thrombosis. Along with sodium salicylate (sc-3520), aspirin has been shown to block the long terminal repeat from the human immunodeficiency virus and the Ig κ enhancer from NFκB dependent transcription.

Solubility
Soluble in 100% ethanol (80mg/ml), DMSO (41mg/ml) or dimethyl formamide (30mg/ml); slightly soluble in Phosphate-buffered saline (PBS), pH 7.2 (2.7mg/ml).

Notes
Store at room temperature. Ensure good ventilation.
Specifications

Specifications

O-Acetylsalicylic acid
133°C to 140°C
250°C (482°F)
MFCD00002430
779271
aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
BSYNRYMUTXBXSQ-UHFFFAOYSA-N
2-acetyloxybenzoic acid
2244
180.15
50-78-2
1.4
C9H8O4
2500 g
14,851
Soluble in 100% ethanol (80mg/ml),DMSO (41mg/ml) or dimethyl formamide (30mg/ml); slightly soluble in Phosphate-buffered saline (PBS),pH 7.2 (2.7mg/ml).
CC(=O)OC1=CC=CC=C1C(O)=O
180.16
CHEBI:15365
99%

Chemical Identifiers

50-78-2
180.16
BSYNRYMUTXBXSQ-UHFFFAOYSA-N
2244
2-acetyloxybenzoic acid
C9H8O4
MFCD00002430
aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
CHEBI:15365
CC(=O)OC1=CC=CC=C1C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302-H500

EINECSNumber : 200-064-1

RTECSNumber : VO0700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only