missing translation for 'onlineSavingsMsg'
Learn More

(-)-O-Acetyl-L-malic Anhydride 97.0+%, TCI America™

Catalog No. A11015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11015G 5 g
A110125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11015G Supplier TCI America Supplier No. A11015G
Only null left

Chemical Identifiers

CAS 59025-03-5
Molecular Formula C6H6O5
Molecular Weight (g/mol) 158.109
MDL Number MFCD00082708
InChI Key SSWJHSASZZAIAU-BYPYZUCNSA-N
Synonym s---2-acetoxysuccinic anhydride, --o-acetyl-l-malic anhydride, s-2-acetoxysuccinic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, 3s, s-acetoxysuccinic anhydride, --o-acetylmalic anhydride, o-acetyl-l-malic anhydride, s-3-acetoxy-dihydro-2,5-furandione, 2,5-dioxotetrahydro-3-furanyl acetate #, s-2,5-dioxotetrahydrofuran-3-yl acetate
PubChem CID 11126528
IUPAC Name [(3S)-2,5-dioxooxolan-3-yl] acetate
SMILES CC(=O)OC1CC(=O)OC1=O

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
Formula Weight 158.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-O-Acetyl-L-malic Anhydride