missing translation for 'onlineSavingsMsg'
Learn More

O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. D32635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32635G 5 g
D326325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32635G Supplier TCI America Supplier No. D32635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 125700-69-8
Molecular Formula C12H16BF4N5O2
Molecular Weight (g/mol) 349.10
MDL Number MFCD00077730
InChI Key FOBCPCIJLQTYBT-UHFFFAOYSA-N
Synonym tdbtu, n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluronium tetrafluoroborate, 1,1,3,3-tetramethyl-2-4-oxobenzo d 1,2,3 triazin-3 4h-yl uronium tetrafluoroborate, o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl-n,n,n',n'-tetramethyluroniumtetrafluoroborate, tdbtu; n,n,n',n'-tetramethyl-o-3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl uronium tetrafluoroborate, dimethylamino 4-oxo-1,2,3-benzotriazin-3-yl oxy methylidene dimethylazanium tetrafluoroborate, pubchem12760, acmc-1cac0, d04fkx
PubChem CID 10882602
IUPAC Name [(dimethylamino)[(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=CC=C2C1=O)=[N+](C)C

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 349.10
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N′,N′-tetramethyluronium Tetrafluorobo
TSCA No
Recommended Storage Refrigerator