Learn More
Thermo Scientific™ Nunclon™ Sphera™ Flasks
Catalog No. 12566440
Change view
Click to view available options
Culture Area:
25 cm²
75 cm²
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area
12566440 75 cm²
12566439 25 cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12566440 Supplier Thermo Scientific™ Supplier No. 174952
Only null left
Features a superior cell culture surface with demonstrated performance that elevates stem cell and cell culture research.
With the super low cell attachment surface, Thermo Scientific™ Nunclon™ Sphera™ Flasks consistently promote 3D spheroids and organoids culture, aiding you in cancer spheroid formation and stem cell differentiation for disease modeling and drug screening.
Significant qualities of the Nunclon Sphera culture surface:
Optimizing spheroid formation
  • Allows cells to grow in suspension with virtually no cell attachment
  • Supports many different cell types and their ability to generate spheroids
  • Enables consistent, repeatable cell growth
Growing 3D cancer cultures
  • Minimizes variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumor growth, providing a better model system for studying tumor cell progression and efficacy of anticancer agents in vitro
Promoting formation of embryoid bodies (EB)
  • Demonstrates superior quality for embryoid body formation of pluripotent stem cells with minimal spontaneous differentiation
  • Resultant cells able to differentiate into all three germ layers
Specifications
Certifications/Compliance Animal origin-free; nonpyrogenic, endotoxin <0.5 EU/mL; USP biological reactivity class VI; SAL 10-6
Description T75 Nunclon Sphera EasYFlask, case of 24
No. per Case 24
No. per Pack 4
Surface Coating Nunclon Sphera
Sterility Sterile
Closure Type Filter
Culture Area 75 cm²