Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Catalog No. 1256566
Change view
Click to view available options
No. of Wells:
96
No. per Pack:
1 Microplate
10 Microplates
No. per Case:
160 Microplates
50 Microplates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells No. per Pack No. per Case
1256566 96 1 Microplate 50 Microplates
12565383 96 10 Microplates 160 Microplates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1256566 Supplier Thermo Scientific™ Supplier No. 167008
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Color: Clear; Sterility: Sterile; Lid: With Lid
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance ISO 9001:2008, ISO 13485:2003
Certified Non-pyrogenic
Format Standard
Lid With Lid
Description 96F well plate, Nunclon Delta, clear, with lid, Sterile (Pack of 1, Case of 50)
Array 8 x 12
Autoclavable Not Autoclavable
Packaging 1/Pk., 50/Cs.
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Nunclon Delta
Well Shape Flat Bottom (F-well)
No. per Case 50 Microplates
No. per Pack 1 Microplate
Quantity 50/Cs.
Show More Show Less