missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Disposable Plastic Centrifuge Tubes

Numéro de catalogue. 12565201
Modifier l'affichage
Click to view available options
Capacity (Metric):
5 mL
12 mL
Bottom Shape:
Round
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Capacity (Metric) Bottom Shape Sterility
12565201 12 mL Round Sterile
12565202 12 mL Round Non-sterile
12565239 5 mL Round Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 12565201 Fournisseur Thermo Scientific™ Code fournisseur 343141
Il en reste null

Maintain a high level of accuracy while increasing lab safety with these disposable plastic centrifuge tubes, which are an excellent alternative to glass.

Maintain a high level of accuracy while increasing lab safety with Thermo Scientific™ Nunc™ Disposable Plastic Centrifuge Tubes, which are an excellent alternative to glass. Selected tubes are relative centrifugal force (RCF) tested. Nunc Labware Products are made from high purity resins and molded using our state of the art processes.

Features:

  • Polyethylene (PE) and polypropylene (PP) tubes are particularly useful in serology where the low adsorption of proteins is of great importance
  • Polystyrene (PS) tubes are transparent and suitable for a broad range of applications
  • Polyethylene and polypropylene tubes have good chemical resistance and high mechanical strength
  • Different cap types are available for a variety of applications and fit most standard centrifuges

Includes:

Cat. nos. 343141, 345608 supplied with mounted stoppers for sterile work.

Spécifications

Capacity (Metric) 12 mL
Disposable Single-use
Material Polystyrene
No. per Case 3600
Sterility Sterile
Shape Round
Bottom Shape Round
Graduations 5 and 10 mL
Product Type Centrifuge Tube
Max. RCF 3000 x g