Learn More
Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates.
Supplier: Thermo Scientific™ 268152
Description
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates. The clear and U shaped wells are ideal for solution-based assays (e.g. agglutination assays) and common serial dilutions, allowing clean and easy pipetting. Available in a variety of options for surfaces, sterility, closures, and packaging sizes.
- Clear polystyrene has excellent optical properties ideal for optical measurements
- Thermo Scientific™ Nunclon™ Delta is a hydrophilic surface treatment whereas the non-treated polystyrene is more hydrophobic
- Customized bar coding is available
Warranty: 90 days
Chemical Identifiers
| Standard | |
| Nunc | |
| Polystyrene | |
| Non-treated |
| 8 x 12 | |
| Microplate | |
| 96 | |
| Round Bottom (U-well) |
Specifications
| American National Standards (ANSI) (ANSI/SBS 1-2004) | |
| Without Lid | |
| 10 Pk. | |
| Clear | |
| Polystyrene | |
| Non-sterile | |
| Round Bottom (U-well) | |
| 10 |
| Standard | |
| 8 x 12 | |
| Nunc | |
| Microplate | |
| 96 | |
| Non-treated | |
| 180 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.