missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 1.8mL Externally-Threaded Universal Tubes and Racks

Numéro de catalogue. p-6020016
Modifier l'affichage
Click to view available options
Barcode:
2D on Tube Bottom
2D on Tube Bottom, Linear on Tube Side
2D on Tube, 2D and Linear on Rack
None
Packaging:
10 Bags of 48 Tubes
10 Racks of 48 Tubes
Bulk
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Barcode Packaging
12590005 2D on Tube Bottom 10 Bags of 48 Tubes
12590006 - -
12590007 2D on Tube Bottom Bulk
12565300 2D on Tube Bottom, Linear on Tube Side Bulk
12590008 None Bulk
12570239 2D on Tube, 2D and Linear on Rack 10 Racks of 48 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. 12590005 Fournisseur Thermo Scientific™ Code fournisseur 374500
Il en reste null

Use the automation friendly tubes to improve the efficiency and throughput of your sample storage workflow. The latch rack offers ANSI SBS footprint compatible with automation platforms for liquid handling and sample storage. Tubes are available racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.

Use the automation friendly Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes to improve the efficiency and throughput of your sample storage workflow. The latch rack offers ANSI SBS footprint compatible with automation platforms for liquid handling and sample storage. Tubes are available racked or in bulk, with 2D barcodes or non-coded. Empty racks are also available.

Scalable

  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cyrostorage boxes made for 10x10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack

Innovative Universal Cap Design

  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen

Secure Tracking

  • Permanently-bonded,unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 10superscript 6 and is made with class VI medical grade resins

Spécifications

Color Natural
Sterility Sterile SAL 10-6
Product Type Externally-threaded Universal Tube
Description Nunc Universal tubes with 2D code in latch rack
Closure Color Natural
Capacity (Metric) 1.8 mL
Bottom Shape Flat
Material Plastic
Thread Style External
For Use With (Equipment) Cryobank
Includes White patch and graduations
No. per Case 480/Cs.
No. per Pack 48/Pk.
Packaging 10 Bags of 48 Tubes
Barcode 2D on Tube Bottom
Afficher plus de résultats Afficher moins de résultats