Learn More
Syngene NuGenius XE
Catalog No. 01001819
Change view
Click to view available options
For Use With (Application):
Gel Imaging
Catalog No. For Use With (Application)
01001819 Gel Imaging
1 options
N/A
Catalog No. 01001819 Supplier Syngene Supplier No. NUGENIUSXE
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
N/A
Compact gel imaging system for imaging DNA/RNA, stain free, visible protein gels and autorads. With a built-in PC and intuitive touchscreen.
Compact gel imaging system for imaging DNA/RNA, stain free, visible protein gels and autorads. With a built-in PC and intuitive touchscreen.
  • The NuGenius XE system is a powerful system with a groundbreaking 5M pixel camera that gives exquisite pixel resolution and unrivalled sensitivity in its class.
  • The system uses a F1.2 motor driven lens to enable perfect gel imaging.
  • The maximum viewing area is 20 x 24cm, which is very large for such a small, compact unit.
  • A flexible system with internal lighting including a UV transilluminator option for working with DNA gels, stain free and autorads.
  • The overhead LED white lighting is included as standard for easy sample positioning and focusing.
  • The NuGenius XE is a versatile system that has been designed for stain free applications. Its camera enables the sample to be exposed for extended periods.
  • Safely excise your DNA bands from agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
  • An intuitive system that permits images to be saved directly to USB or network for easy transfer.
  • Quickly generate quantitative data with GeneTools analysis software.
  • NuGenius XE comes with unlimited copies of GeneTools.
Specifications
For Use With (Application) Gel Imaging
Product Type Gel Imaging System
Display Type LED
N/A