missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Nucleon™ PhytoPure™ Genomic DNA Extraction Kits
Nucleon PhytoPure DNA Extraction kits use a phytopure resin to bind plant polysaccharides ensuring high yields of high-quality plant genomic DNA in only one hour
$409.57 - $798.49
Specifications
| Description | Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well |
|---|---|
| Sample Type | Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried) |
| For Use With (Application) | Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material |
| Includes | Reagents 1 and 2 and PhytoPure™ Resin |
Includes: Reagents 1 and 2 and PhytoPure Resin
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
45-000-958
|
Cytiva
RPN8510 |
100 mg |
Each for $409.57
|
|
|||||
|
45-000-959
|
Cytiva
RPN8511 |
1000 mg |
Each for $798.49
|
|
|||||
Description
- Rapid protocol does not require phenol or CTAB
- Two kit sizes provide economical scale-up using the same phenol-free protocol
- Effective removal of polysaccharides minimises inhibition of downstream applications
Specifications
| Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well | |
| Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried) | |
| Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material | |
| Reagents 1 and 2 and PhytoPure™ Resin |