Learn More
Cytiva Nucleon™ PhytoPure™ Genomic DNA Extraction Kits
Nucleon PhytoPure DNA Extraction kits use a phytopure resin to bind plant polysaccharides ensuring high yields of high-quality plant genomic DNA in only one hour
Supplier: Cytiva RPN8510
Description
- Rapid protocol does not require phenol or CTAB
- Two kit sizes provide economical scale-up using the same phenol-free protocol
- Effective removal of polysaccharides minimises inhibition of downstream applications
Specifications
| Quality, length, and purity of a phenol-chloroform extraction without phenol. Nucleon PhytoPure enables rapid chloroform extraction of high-quality, high molecular weight genomic DNA from plant and fungal samples with efficient removal of protein as well | |
| Successfully extracts DNA from many plant species, starting with fresh, frozen, or freeze dried material | |
| illustra Nucleon PhytoPure |
| Barley (Fresh, Frozen or Freeze Dried), Maize (Fresh, Frozen or Freeze Dried), Rye (Fresh, Frozen or Freeze Dried), Wheat (Fresh, Frozen or Freeze Dried) | |
| Reagents 1 and 2 and PhytoPure™ Resin | |
| 100 mg |