Cell-ID Intercalator-103Rh is a cationic nucleic acid intercalator that contains natural-abundance rhodium ( 103Rh) and is used in CyTOF® analysis for discrimination of dead cells from live cells (if staining is done before cell fixation) or to discriminate single nucleated cells from doublets (if staining is done post-fixation). It is a live cell membrane-impermeable dye. For CyTOF® 2 and Helios™ instruments we recommend using Cell-ID Intercalator-Ir (201192B) for nucleated cell discrimination and Cisplatin-194 (201194) or -198 (201198) for dead cell discrimination.