missing translation for 'onlineSavingsMsg'
Learn More

Innovatek NRSII™ Transwab™ Environmental Swabs

Catalog No. 361041858
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
361041858 1 mL
361041856 5 mL
361041857 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 361041858 Supplier Innovatek Supplier No. 4080MW786

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Innovatek NRSII™ Transwab™ Environmental Swabs accurately recover residual bacteria on food contact surfaces

  • Swab is preimmersed, ready for use with a self standing tube
  • High visibility blue cap and swab is easy to find and retrieve if accidentally dropped
  • Conforms to ISO 18593 and other national and international standards
  • Neutralisers allow accurate recovery of residual bacteria on food contact surfaces
  • Precise fill for accurate assessment of contamination levels
  • Reduced packaging & storage space
  • Ambient storage and transport

Specifications

Media Type Neutralizers
Certifications/Compliance ISO 18593
Closure Color High Visibility Blue
Color Clear
For Use With (Application) Sampling Previously Cleaned Surfaces, Food Safety
Quantity 50/Pk.
Storage Requirements Ambient Conditions
Volume (Metric) 1 mL