missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ROR alpha Plasmid

Catalog No. NBP231076
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231076 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP231076 Supplier R&D Systems™ Supplier No. NBP231076

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. For expression of hROR alpha protein in mammalian cells, induction of G6Pase promoter activity

The ROR alpha Plasmidis a mammalian expression vector construct, pCMV/hRORalpha, containing coding sequence of human RAR-related orphan receptor alpha.

  • Species: Human
  • Immunogen: ROR alpha Plasmid Expression of hROR alpha protein in mammalian cells. Induction of G6Pase promoter activity
  • Total contents of 10μg is enough for one 96-well cell transfection

Recommended Storage

Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.

Specifications

Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Product Line Novus
Product Type ROR alpha Plasmid
Quantity 10 μg
Synonym hROR, hROR alpha ROR, ROR alpha