missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Ramos Cell Lysate
Description
The Ramos Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, Immunohistochemistry.
Specifications
Specifications
| Cell Line | Ramos cell line lysate (human Lymphoblast, Burkitt's lymphoma) |
| Quantity | 0.1 mg |
| Host Species | Human |
| Content And Storage | -80°C in the dark. Avoid freeze-thaw cycles. |
| Product Line | Novus |
For Research Use Only