missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HEK293 Cell Lysate

Catalog No. NBP225048
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225048 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225048 Supplier Novus Biologicals™ Supplier No. NBP225048

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Applications: SDS-Page

The HEK293 Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.

Specifications

Quantity 0.1 mg
Storage Buffer 1X SDS sample buffer: 60mM Tris-HCl (pH 6.8) 10mM EDTA 2 % SDS 0.03% Bromophenol Blue 10% glycerol 0.3% Beta-mercaptoethanol
Test Specificity This lysate was prepared from a cell culture of HEK 293 (human embryonic kidney cells).
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.

For Research Use Only