missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Power Supply Adapters

Catalog No. ZA10001
Encompass
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ZA10001 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZA10001 Supplier Invitrogen™ Supplier No. ZA10001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One set of adapters and one Allen wrench

The Novex Power Supply Adapters allow users to connect the covered power leads of electrophoresis units to certain power supplies.

The Novex Power Supply Adapters allow you to connect the covered power leads of electrophoresis units to certain power supplies (for example, some power supplies from Bio-Rad Laboratories, Thermo Fisher Scientific).

The following electrophoresis units from Invitrogen have covered power leads to be in compliance with the latest IEC (International Electrotechnical Commission) and EN (European) standards, and reduce the risk of electrical shock due to exposure to bare metal.

• ZOOM IPGRunner Mini-Cell and ZOOM IEF Fractionator
• XCell4 SureLock Midi-Cell
• Mini Gel Tank
• XCell SureLock Mini-Cell (has retractable power leads)

If you are using any of the above electrophoresis units with power supplies that are not designed for use with covered or retractable power leads, you need to use Novex Power Supply Adapters.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Contains 1 set of adapters and 1 Allen wrench
For Use With (Equipment) XCell SureLock Mini, ZOOM IPGRunner Mini, XCell4 SureLock Midi, ZOOM IEF Fractionator, Mini Gel Tank
Product Line Novex
Quantity 1 set
Shipping Condition Room Temperature
Can I purchase replacement parts for the Mini Blot Module?

Here are the replacement parts we offer for the Mini Blot Module:

- Mini Blot Module Gasket, Cat. No. B1001
- Gel Knife, Cat. No. EI9010
- Bolt Tank Base, Cat. No. B4478640
- Gel Runner Tank, Cat. No. B4478641
- Mini Gel Tank Lid, Cat. No. A25944
- Cassette Clamp, Right, Cat. No. B4478592
- Cassette Clamp, Left, Cat. No. B4478593
- Power Supply Adapters, Cat. No. ZA10001

Can I purchase replacement parts for the XCell4 SureLock Midi-Cell?

Here are the replacement parts we offer for the XCell4 SureLock Midi-Cell. They can only be ordered by phone:

- XCell4 SureLock Midi-Cell Lid: Cat. No. WR0101
- XCell4 SureLock Midi-Cell Lower Buffer Chamber: Cat. No. WR0102
- XCell4 SureLock Midi-Cell Displacement Dam: Cat. No. WR0106
- XCell SureLock Upgrade kit: Cat. No. WR0112 (contains 1 SureLock Assembly, 1 buffer dam, and 2 cores) Please note that in addition to ordering on the phone by calling Customer Service, this item can also be ordered using the “Quick Order” link (https://www.thermofisher.com/store/quick-order) that can be found at the top right of most pages on our website. The price will display when the item is added to your shopping cart.

What are the components of the XCell4 SureLock Midi-Cell?

The XCell4 SureLock Midi-Cell includes the lower buffer chamber, lid with cables, two buffer cores, buffer dam, displacement dam, SureLock assembly, and gel knife. For descriptions and images of the various parts, please refer to pages 1-3 of the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/xcell4_midicell_man.pdf).

Can I purchase replacement parts for the XCell SureLock Mini-Cell?

Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052

What are the components of the XCell SureLock Mini-Cell?

The XCell SureLock Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.

I am running my Tris-Glycine gel using the XCell SureLock Mini Cell but the leads do not fit into my power supply. Can you please help?

You may purchase the ZOOM adapters, Cat. No. ZA10001 to help you connect your leads to the power supply


For Research Use Only. Not for use in diagnostic procedures.