missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ni-NTA Agarose

Catalog No. R90115
Encompass
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R90115 25 mL
R90101 10 mL
R90110 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R90115 Supplier Invitrogen™ Supplier No. R90115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

The Ni-NTA resin is pre-charged and able to bind up to 50mg of recombinant protein per 1mL of resin. It is provided as a 50% slurry in 30% ethanol. The resin will appear blue in color when charged with Ni2+

Nickel-charged affinity resin that can be used to purify recombinant proteins containing a polyhistidine (6xHis) sequence

  • Ni-NTA Agarose is a nickel-charged affinity resin that can be used to purify recombinant proteins containing a polyhistidine (6xHis) sequence
  • Proteins bound to the resin may be eluted with either low pH buffer or by competition with imidazole or histidine
  • One-step purification can be performed under both native and denaturing conditions
  • Ni-NTA uses the chelating ligand nitrilotriacetic acid (NTA) coupled to a cross-linked 6% agarose resin that is suitable for use in batch and gravity flow applications
  • Guaranteed stable for 6 months when properly stored at 4°C

Specifications

Content And Storage The Ni-NTA resin is pre-charged and able to bind up to 50 mg of recombinant protein per 1 ml of resin. It is provided as a 50% slurry in 30% ethanol. The resin will appear blue in color when charged with Ni2+ . Store at +4°C. Ni-NTA resin is guaranteed stable for 6 months when properly stored.
Stationary Phase Ni-NTA
Type Agarose
Quantity 25 mL
Form Suspension
Column Type Affinity Column
Can ProBond or Ni-NTA beads be used for large-scale preparations?

ProBond and Ni-NTA beads can be used in FPLC columns. However, the beads can only withstand low pressure (~43.5 psi max).

What should the typical protein recovery be when using the Probond Purification System or Ni-NTA Purification system?

Both systems are qualified by purifying 2 mg of myoglobin protein on a column and performing a Bradford assay. Protein recovery must be 75% or higher.

For Research Use Only. Not for use in diagnostic procedures.