missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ IEF Cathode Buffer pH 3-7 (10X)

Catalog No. LC5370
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC5370 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC5370 Supplier Invitrogen™ Supplier No. LC5370

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimized pre-mixed IEF buffers reduce variability and ensure consistent results

Novex IEF Cathode Buffer pH 3-7 (10X) is optimized pre-mixed IEF cathod buffer for using with Novex pH 3-7 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.

Isoelectric Focusing, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in refrigerator (2–8°C).
Buffer Anode & Cathode Buffers
Concentration 10 X
Gel Type IEF
pH Range 3 to 7
Gel Compatibility Novex™ IEF Gels
Quantity 125 mL
Shipping Condition Wet Ice
Product Line Novex
Product Type IEF Cathode Buffer
When staining IEF gels with the Colloidal Blue Staining Kit, is it necessary to use the fixing solution?

Yes, the fix serves two purposes: it fixes the sample in the IEF gel and it helps to remove gel background.

If you do not use the fixing solution, the background on the gels will be high and detection will be less sensitive.

High background is caused by ampholytes remaining in the gel.

When running an IEF gel, is it okay to use just arginine, instead of a mix of arginine and lysine, in the cathode buffer? Does it matter whether the D, L or D/L form of arginine is used?

Yes, you can use just arginine (either form of arginine) as long as the arginine free base form is used and not the hydrochloric form. Note that the lysine used to make the mixed buffer should be free base lysine; free acid lysine should not be used as a substitute. The reason for adding basic amino acids to the cathode buffer is to increase the ionic strength of the buffer in order to fortify the basic end.

For Research Use Only. Not for use in diagnostic procedures.