missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Antibody Coupling Kit

Catalog No. 14311D
Encompass
Change view
Click to view available options
Quantity:
60 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14311D 60 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14311D Supplier Invitrogen™ Supplier No. 14311D

Includes

60mg Dynabeads M-270 Epoxy and Buffers required for covalent coupling. Dynabeads M-270 Epoxy supplied freeze dried. Multiple buffers provided in various volumes.

Requires

Antibody

The Dynabeads™ Antibody Coupling Kit is intended for the coupling of an antibody or protein to the surface of Dynabeads™ M-270 Epoxy beads.

The Dynabeads™ Antibody Coupling Kit is intended for the coupling of an antibody or protein to the surface of Dynabeads™ M-270 Epoxy beads. The coupled beads and supplied buffers can then be used for experiments such as immunoassays, immunoprecipitation (IP), Co-IP of protein complexes, chromatin IP (ChIP), RNA-IP (RIP), cell isolation, etc. The Dynabeads Epoxy beads in this kit do not contain Tween™ detergent making them suitable for mass spectrometry (MS) analysis. The kit includes the buffers needed for efficient antibody coupling and washing steps. Note that the SB buffer supplied in the kit does contain Tween™ detergent, which means the buffer will need to be replaced with standard TBS or PBS buffer if the kit is to be used for MS.

Benefits of Dynabeads Antibody Coupling Kit include:
Convenience—contains optimized buffers and epoxy-activated Dynabeads magnetic beads
Efficiency­—easy-to-use protocol with minimal hands-on time
Flexibility­—allows coupling of any antibody of your choice
Robustness—covalent antibody coupling to the beads avoids co-elution of the antibody with the target proteins

Efficient and easy coupling
With this kit antibodies are covalently coupled to Dynabeads M-270 Epoxy beads, minimizing the risk of bound antibodies contaminating the final eluate. Dynabeads M-270 Epoxy beads exhibit extremely low background binding and do not require blocking before use. Other proteins such as lectins and enzymes may also be coupled using the same beads, buffers, and protocol (depending on the stability and functionality of the protein/ligand you are using).

Magnetic bead-based separation offers easy handling
Captured proteins and protein complexes are easily separated, washed, and eluted using a DynaMag magnet due to the magnetic properties of the Dynabeads M-270 Epoxy beads. The supplied buffers are optimized to give consistent and reliable results. In addition to Dynabeads M-270 Epoxy beads, the kit includes five different binding, washing, and elution buffers. Target specific antibody is required for coupling, but not included.

Automation-ready for use with KingFisher purification systems
The 2.8-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems or liquid handlers.

Commercial supply
Our manufacturing sites are ISO 13485-certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads Antibody Coupling Kit in a commercial product or service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or read more on our Dynabeads OEM page.

Order Info

Shipping Condition: Room Temperature

Specifications

Description For immoblization of antibodies to magnetic beads
Material Polystyrene
Content And Storage Dynabeads M-270 Epoxy beads (60 mg) and 5 different buffers for covalent coupling and washing Store at 2°C to 8°C.

Diameter (Metric) 2.8 μm
For Use With (Equipment) KingFisher™ Sample Purification Systems, DynaMag™ Magnets
High-throughput Compatibility High-throughput Compatible
Shipping Condition Ambient temperature
Sample Type Intact Proteins (antibodies, purified antibodies, peptides, functional enzymes)
Surface Functionality Epoxy
Quantity 60 mg
For Use With (Application) Immunoprecipitation
Type Antibody Coupling Kit
Final Product Type Antibody (or other protein ligand) coupled Dynabeads
Format Freeze-dried beads
Color Brown
Certifications/Compliance ISO9001 and ISO13485
Regulatory Status RUO
Uniformity Monosized 2.8 μm (CV <5%)
Show More Show Less
My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

What sizes do you offer for the Dynabeads magnetic beads?

Dynabeads magnetic beads come in three sizes: 4.5 µm (M-450), 2.8 µm (M-270/M-280), and 1 µm (MyOne beads). The largest of the Dynabeads magnetic beads is ideal for big targets like cells. The 2.8 µm beads are recommended for proteomics and molecular applications. The smallest of the beads, 1 µm, are ideal for automated handling.

Can Dynabeads magnetic beads be sonicated?

In general, short sonication is a good way to reduce aggregation of the beads and ensure optimal homogenous conditions at the time of ligand addition when coating the beads. When target is bound to the beads, more care is needed, as the binding might break. The streptavidin beads themselves should tolerate sonication. We have not tested sonication for long periods, but 5 minutes is fine. We do not have information about the streptavidin-biotin interaction being broken by such treatment.

Can Dynabeads magnetic beads be sterilized?

If desired, the uncoated epoxy or tosylactivated beads can be sterilized by washing with 70% ethanol. Coated beads cannot be sterilized.

What are Dynabeads magnetic beads?

Dynabeads magnetic beads are uniform, non-porous, superparamagnetic, monodispersed and highly cross-linked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The magnetic material within the Dynabeads magnetic beads consists of a mixture of maghemite (gamma-Fe2O3) and magnetite (Fe3O4). The iron content (Fe) of the beads is 12% by weight in Dynabeads magnetic beads M-280 and 20% by weight in Dynabeads magnetic beads M-450. The Dynabeads magnetic beads are coated with a thin polystyrene shell which encases the magnetic material, and prevents any leakage from the beads or trapping of ligands in the bead interior. The shell also protects the target from exposure to iron while providing a defined surface area for the adsorption or coupling of various molecules.

Uniformity of bead size and shape provides consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.

The Dynabeads magnetic beads are available in three different sizes: 4.5 µm (M-450 beads), 2.8 µm (M-270/M-280 beads) and 1 µm (MyOne beads). as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Can I couple a large amount of antibody to Dynabeads magnetic beads using the Dynabeads Antibody Coupling Kit?

Coupling of large amounts of antibody (20 to 30 µg antibody/mg beads) is difficult. If loading 20 µg of antibody to the beads, the total amount of antibody covalently coupled to the beads will not be more than 13 µg/mg beads. Up to 10 µg, you will find a linear relationship between added antibody and coupled antibody. Above these concentrations, the antibody will precipitate down to the bead surface but a reduction in covalent bonds will be observed.

What should I do when the antibody volume equals or exceeds the C1 volume when using the Dynabeads Antibody Coupling Kit?

If the volume of antibody is equal to (or exceeds) the volume of C1, the antibody solution can be used directly (as long as the pH is approx. 7.4). So, for 5 mg Dynabeads magnetic beads, add 250 µL antibody + 250 µL C2.

Can I couple more than one ligand to the Dynabeads magnetic beads?

Yes, but this would require some optimization of the coupling process since the two ligands may have inherent differences in hydrophobic properties and isoelectric points that would in turn influence the coupling conditions (pH, temperature, etc.). If a particular ratio of the two ligands on the Dynabeads magnetic beads surface is desired, one would have to test the binding capacity for each ligand separately and then optimize the coupling process for the combination of ligands. For coupling more than one ligand, we recommend the Dynabeads M-270 Epoxy magnetic beads - either the stand-alone beads, Cat. No. 14301 (coupling buffers are not included) or Dynabeads Antibody Coupling kit, Cat. No. 14311D (coupling buffers are included) as these beads are used in our Dynabeads Human T-Activator CD3/CD28 magnetic beads and Dynabeads Human T-Activator CD3/CD28/CD137 magnetic beads where 2 and 3 antibodies are coupled to the beads in certain ratios.

How can antibodies be coupled to Dynabeads magnetic beads?

There are two main ways of coupling antibodies to the beads (with several different options within the two groups): Covalent coupling and Noncovalent coupling.

For covalent coupling, there are several choices including:
Dynabeads M-280 Tosylactivated magnetic beads (Cat. No. 14203)
Dynabeads M-270 Epoxy magnetic beads (Cat. No. 14301)
Dynabeads Antibody Coupling Kit (Cat. No. 14311D, Dynabeads M-270 Epoxy and buffers for covalent coupling)

For co-immunoprecipitation, Dynabeads Co-Immunoprecipitation Kit (Cat. No. 143-21D, Dynabeads M-270 Epoxy magnetic beads, buffers for covalent coupling, and buffers optimized for co-immunoprecipitation; see Alber F et al. (2007) Determining the architectures of macromolecular assemblies. Nature 450, 683-694).

Dynabeads magnetic beads can also be used for immunoprecipitation, which relies on noncovalent binding of antibodies. The most common products used for noncovalent binding of antibodies to Dynabeads magnetic beads are:
Dynabeads Protein A magnetic beads (Cat. No. 10001D)
Dynabeads Protein G magnetic beads (Cat. No. 10003D)
Immunoprecipitation Kit - Dynabeads Protein A magnetic beads (Cat. No. 10006, containing Dynabeads magnetic beads and optimized buffers for immunoprecipitation)
Immunoprecipitation Kit - Dynabeads Protein G magnetic beads (Cat. No. 10007, Dynabeads magnetic beads and optimized buffers for immunoprecipitation)
Dynabeads magnetic beads coated with secondary antibodies. (Dynabeads Pan Mouse IgG magnetic beads (Cat. No. 11041) are 4.5 µm beads developed for cell separation that have increased capacity per volume when using Dynabeads M-280 Sheep Anti-Mouse IgG magnetic beads.

Thus, there are several products that can be considered for performing immunoprecipitation depending on preferences. If the target is the same as heavy or light chain antibody, we recommend covalently binding the antibody to the bead surface. This can be done either by cross-linking the antibody to beads coated with Protein A or G or with secondary antibody or by using one of the surface-activated Dynabeads products. As your primary antibody can be used in combination with Dynabeads Protein G magnetic beads (Cat. No. 100-03D) or Immunoprecipitation Kit - Dynabeads Protein G magnetic beads (Cat. No. 100-07D, contains immunoprecipitation buffers as well) or Dynabeads Sheep Anti-Mouse IgG magnetic beads, using a cross-linker will ensure covalent binding of your primary antibody. Depending on the antibody, the functionality of the antibody can be affected. The other option would be the surface-activated Dynabeads products. This is an easy but more time-consuming approach, since the coupling takes an overnight incubation, but it ensures functional antibodies that are not eluted off during elution. For this approach, we recommend the Dynabeads Antibody Coupling Kit (Cat. No. 14311D, contains both surface-activated beads and optimized buffers for covalent coupling). as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

For how long can Dynabeads magnetic beads be stored?

Storage should be at 2-8 degrees C. Freezing Dynabeads magnetic beads is not recommended. Provided the Dynabeads magnetic beads are stored correctly, quality is guaranteed until the expiration date stated on the label. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

How should I store coated Dynabeads magnetic beads if I want to reuse them?

Dynabeads magnetic beads coated with antibody/ligand may be stored at 2-8 degrees C without loss of antigen binding capacity. For long-term storage, a final concentration of 0.02% NaN3 may be added to the antibody-coupled beads in a physiological buffer. Please note that not all coupled antibodies retain their function in long term storage. Verify your coupled antibody stability by testing in small scale. After storage, coated Dynabeads magnetic beads should be washed once in PBS/BSA for 5 min before use.

How can I dissociate isolated proteins from Dynabeads magnetic beads?

Standard elution methods are used to dissociate the isolated protein from Dynabeads magnetic beads. The most suitable elution method depends on the characteristics of the isolated protein and the desired downstream application of the eluted protein. In many cases, Dynabeads magnetic beads can be recovered for reuse after elution. Some standard elution methods are listed below.

-pH change: elution can be achieved by reducing pH (for example, by using 0.1 M citrate (pH 2-3) as the elution buffer). Dynabeads magnetic beads are stable between pH 4-13. If the Dynabeads magnetic beads are exposed for a prolonged period of time to pH below 4, the beads may be adversely affected.

-Change of ionic strength: high-salt concentration buffers (e.g., NaI, KI, MgCl, KCl) can be used to elute isolated proteins. Optimization is required, by step-wise elution starting at 1 M and increasing to 3 M.

-Affinity elution: with this method, the eluting agent competes for the binding of the protein or the binding of the ligand e.g., elution of glycoproteins from a lectin coupled to Dynabeads magnetic beads may be achieved by the addition of the free sugar.

-Denaturing eluents: as a last resort, denaturing eluents such as chaotropic salts may be used to alter the structure of the protein. The proteins on the bead surface and the eluted proteins will be denatured.

-Polarity reducing agents : substances that reduce the polarity of the buffer often disrupt the hydrophobic interactions between antibody and protein. Dioxane or ethylene glycol may be used to reduce the polarity of the eluent. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What is the definition of superparamagnetic, and what does this mean for my cell isolation application with Dynabeads magnetic beads?

Superparamagnetic means that the Dynabeads magnetic beads exhibit magnetic properties when placed within a magnetic field, but have no residual magnetism when removed from the magnetic field.

This means that your targeted cells, proteins, or nucleic acids are only subjected to magnetic forces during the time the beads are on the magnet. The beads do not aggregate, but remain evenly dispersed in suspension.

Are the antibodies on your Dynabeads magnetic beads for cell isolation/activation/expansion covalently bound to the beads?

Yes. The antibodies are covalently bound and should be very stable in your applications.

What is the average shelf life of Dynabeads magnetic beads?

Depending on the antibody coated on the Dynabeads magnetic beads, the shelf life can vary from 24-36 months.
Some kits may have 18 months shelf life depending on other components supplied in the kit. The kits are guaranteed for 6 months from when you receive them. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

I was able to immunoprecipitate my protein using Dynabeads magnetic beads before, but after crosslinking the antibody to the beads I get no protein bands on my gel. What could be the problem?

The binding sites of your antibody have likely been altered by the crosslinking. When this occurs, your antibody will show reduced affinity or no affinity to its target antigen. Another consequence of crosslinking can also be increased affinity for unintended (nonspecific) targets. This is always a high risk with crosslinking, and it is a problem easily avoided by choosing another path to covalent antibody coupling. You can try using the Dynabeads Antibody Coupling Kit. This kit is a far superior solution for covalent antibody coupling to Dynabeads magnetic beads (compared to crosslinking with Dynabeads Protein A or G magnetic beads). The Dynabeads Antibody Coupling Kit is compatible with almost any antibody. It is designed specifically for covalent antibody coupling to Dynabeads magnetic beads. Unlike crosslinking, the Dynabeads Antibody Coupling Kit will not alter antibody specificity or affinity.

What do the designations M-280, M-270, and MyOne mean on Dynabeads magnetic beads?

The M stands for magnetic. M-280 refers to hydrophobic 2.8 micron beads, while M-270 refers to hydrophilic 2.8 micron beads. MyOne refers to 1 micron beads. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What is the detection limit when using Dynabeads magnetic beads for immunoprecipitation (IP)?

Answering this question is not straightforward. It will depend on the detection method. When using HRP (horseradish peroxidase)-based detection system or radioactivity in combination with a good antibody, very little target is required. More target is required when using an AP (alkaline phosphatase)-based detection system. When a sensitive detection system is used, detection will most likely be in the nanogram range. In some cases, pictograms of target can be detected.

What is the elution volume when using Dynabeads magnetic beads for immunoprecipitation (IP)?

Within practical limits, the elution volume can be scaled up or down to suit your experiment. However, volumes less than 10 µL become more difficult to work with. In addition, the amount of target is important. If you have a lot of beads with a lot of bound target in a small elution volume, your elution may not be very efficient. Typically, 15-100 µL of beads may be eluted in 30 µL. For efficient recovery of the antigen and/or binding partners, the elution volume should at minimum equal the volume of the beads.

Can I couple more than one ligand to Dynabeads magnetic beads?

Yes, but this would require some optimization of the coupling process since the two ligands may have inherent differences in hydrophobic properties and isoelectric points that would, in turn, influence the coupling conditions (pH, temperature, etc.). If a particular ratio of the two ligands on the Dynabeads magnetic beads surface is desired, you will need to test the binding capacity for each ligand separately and then optimize the coupling process for the combination of ligands. For coupling more than one ligand, we recommend the Dynabeads M-270 Epoxy magnetic beads-either the stand-alone beads, Cat. No. 14301 (coupling buffers are not included) or Dynabeads Antibody Coupling kit, Cat. No. 14311D (coupling buffers are included).

How can I quantify the amount of antibody bound to Dynabeads magnetic beads?

There are several methods to quantify the amount of antibody bound to the beads. The crudest method is to measure the concentration of antibody in the coupling reaction before and after antibody attachment. Either fluorescence measurements or absorbance at 280 nm can be used. Alternatively, you could measure the amount of antibody bound to the beads by fluorescence, chemiluminescence, or radiolabeling detection methods. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

How long should I incubate my antibody with the lysates?

Incubation time will depend on the immunogenicity of the primary antibody and its binding affinity with the specific antigens. For a good primary antibody, 30-40 minutes incubation should work well. If you are working with a poor antibody or a very low-abundance protein, you could try to increase binding by incubating overnight. However, this also increases the chance of background protein binding.

When should I covalently bind the antibody to the Dynabeads surface?

If the target protein has the same molecular weight as the heavy or light chain antibody, we would recommend covalently binding the antibody to the bead surface. This can be done by either crosslinking the antibody to the Dynabeads Protein A or G magnetic beads, or secondary coated beads, or by using one of the surface-activated Dynabeads magnetic beads. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What is the recommended amount of antibody for a coupling reaction?

The recommended amount of antibody for a coupling reaction is typically 5-7 µg antibody/mg Dynabeads magnetic beads.

What are the general advantages of using Dynabeads magnetic beads for protein isolation?

Using Dynabeads magnetic beads for protein isolation provides several advantages:

-Rapid binding kinetics: since the number of beads per volume for Dynabeads is approximately 1,000 times higher than for the same volume of a Sepharose slurry, the probability for Dynabeads magnetic beads to hit the target is far greater.

-Incubation time: due to the rapid binding kinetics, the protocol is usually very short.
-Low background: due to the rapid binding kinetics and the short incubation time, the background is also very low.
-Trapping of impurities: the beads offer no internal volume for binding or trapping of impurities.
-Low antibody consumption: this is because Dynabeads magnetic beads are nonporous, uniform superparamagnetic, monodispersed, highly crosslinked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The beads are coated with a thin layer of a highly crosslinked polystyrene shell that encases the magnetic material and prevents any leakage from the beads or trapping of ligands in the bead interior. The outer layer also provides a defined surface area for the adsorption or coupling of various molecules such as antibodies. Uniformity of bead size and shape provide consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.
-Reproducibility: due to easier practical handling, such as pipetting. No centrifugation steps or preclearing are required.

Are Dynabeads magnetic beads compatible with dithionite, DTT, and EDTA?

No. Not only is dithionite a reducing agent, but the strong affinity of the dithionite ion for bivalent and trivalent metal cations (M2+, M3+) allows it to enhance the solubility of iron, making it a chelating agent. As a result, the iron in the Dynabeads magnetic beads is reduced and pulled out when they are exposed to dithionite. The same is observed if Dynabeads magnetic beads are exposed to DTT and EDTA. With EDTA, we highly recommend checking the minimal amount of EDTA that your specific molecules would tolerate for binding to the Dynabeads, and if it will affect your specific application. For some applications, low concentrations of EDTA can be tolerated by Dynabeads. On the other hand, using 10 mM EDTA with heating affects the binding of biotin molecules to Dynabeads streptavidin. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Are Dynabeads magnetic beads compatible with Urea?

Yes, they are compatible with 6-8 M Urea when used during post-coupling steps.

Are Dynabeads magnetic beads compatible with centrifugation?

Dynabeads magnetic beads, being magnetic in nature are really not designed to be centrifuged. That being said, the beads themselves are compact, as the pores in the polymer matrix are filled with magnetic material and coated with a final outer polymer shell that will further add to the rigidity of the beads. Hence, pressure should theoretically not be a problem for the beads themselves, but the force exerted by the beads on surrounding cells in the pellet may be detrimental to the cells. as well as our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

What are the benefits of using magnetic beads in immunoprecipitation (IP) experiments?

Magnetic beads, unlike agarose beads, are solid and spherical, and antibody binding is limited to the surface of each bead. While magnetic beads do not have the advantage of a porous center to increase the binding capacity, they are significantly smaller than agarose beads (1 to 4 µm), which collectively gives them adequate surface area-to-volume ratios for optimum antibody binding.

High-power magnets are used to localize magnetic beads to the side of the incubation tube and out of the way to enable cell lysate aspiration without the risk of also aspirating immune complexes bound to the beads. Magnetic separation avoids centrifugation, which can break weak antibody-antigen binding and cause loss of target protein.

However, an issue with the use of magnetic beads is that bead size variations may prevent all beads from localizing to the magnet. Additionally, while immunoprecipitation using agarose beads only requires standard laboratory equipment, the use of magnetic beads for immunoprecipitation applications requires high-power magnetic equipment that can be cost-prohibitive. Read more about our Magnetic Immunoprecipitation Products (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/immunoprecipitation.html#products).


For Research Use Only. Not for use in diagnostic procedures.