missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA Assay Buffer (5X)

Numéro de catalogue. DS98200
Encompass
Modifier l'affichage
Click to view available options
Quantité:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
DS98200 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. DS98200 Fournisseur Invitrogen™ Code fournisseur DS98200
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

For use with antibody pairs (ELISA)

Concentrated 5X Assay Buffer for Antibody Pairs is intended to be used as:

  • Blocking reagent for ELISA plates
  • Diluent for ELISA standards and samples
  • Diluent for the detection antibody
  • Diluent for the HRP conjugate

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

Spécifications

Applications ELISA
Quantité 200 mL
Contenu et stockage Buffer. Store at 2-8°C.
Forme Concentrated
Shipping condition (Condition d'expédition) Wet Ice

For Research Use Only. Not for use in diagnostic procedures.