missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Novex™ Value™ Tris-Glycine Mini Protein Gel, 10%, 1.0 mm, 10-well

Catalog No. XV00100PK20
Encompass
Change view
Click to view available options
Quantity:
20 gels (1 box)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
XV00100PK20 20 gels (1 box)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. XV00100PK20 Supplier Invitrogen™ Supplier No. XV00100PK20

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

20 precast gels

Separation Range: 30 to 250 kd; Separation Type: Denaturing, Native; For Use With: XCell SureLock Mini

Features of the Novex Value gels include:
  • Great balance of value and performance
  • Fast run conditions - separate your proteins using constant voltage in less than 60 minutes
  • Improved shelf life - store gels for at least 12 months at 4°C
  • Usable for native and denaturing protein assays

Warranty

Up to 12 months

Specifications

Content And Storage Store in refrigerator (2–8°C).
Shelf life up to 12 months depending on gel type.
Description 10% Value Tris-Glycine Gels, 1.0 mm, 10-well, Box of 20
Gel Percentage 10%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine
Recommended Applications Denaturing, Native
Sample Loading Volume up to 25 μL
Separation of Protein
Separation Range 30 to 250 kDa
Separation Type Denaturing, Native
Wells 10-well
For Use With (Application) Protein Gel Electrophoresis
Material Tris-Glycine Gel
Mode of Separation Molecular Weight
Quantity 20 gels (1 box)
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Storage Requirements Store at 4°C. Shelf life is 12 months.
Thickness 1.0 mm
Well Design 1D Well
Length (Metric) 8 cm
Length (Metric) Gel 8 cm
Width (Metric) 8 cm
Width (Metric) Gel 8 cm
Product Line Novex Value
Product Type Homogeneous Page
For Use With (Equipment) XCell SureLock Mini-Cell
Show More Show Less
I am transferring a Tris-Glycine gel using constant voltage and the current reading is way over the expected starting current. Can you offer some suggestions?

The most common cause of abnormally high current is the transfer buffer. If the transfer buffer is too concentrated, this leads to increased conductivity and current. High current may also occur if Tris-HCl is accidentally substituted for the Tris base required in the transfer buffer. This will again result in low buffer pH and lead to increased conductivity and current and subsequently, overheating. We recommend checking the transfer buffer and its reagent components and re-diluting or remaking the buffer.

For Research Use Only. Not for use in diagnostic procedures.