Learn More
MilliporeSigma™ Novagen™ RedAlert™ 10X Western Blot Stain
Catalog No. 710783
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
710783 50 mL
1 options
Catalog No. 710783 Supplier MilliporeSigma™ Supplier No. 710783
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Visualizes protein transferred to membranes prior to Western blot analysis.
EMD Millipore Novagen™ RedAlert™ 10X Western Blot Stain binds reversibly to proteins on nitrocellulose and PVDF membranes.
  • Stain proteins bound to cellulose acetate
  • Binds positively charged amino groups nonpolar regions of proteins, resulting in reddish-pink bs
  • Applications:
  • Verifies transfer of proteins before immunological detection on nitrocellulose or PVDF membranes
  • Detects blood serum proteins on cellulose acetate media
  • Packaging: Glass Bottle
Specifications
Quantity 50 mL
Product Type Protein Stain