missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ Overnight Express™ Autoinduction System 1

Induces protein expression without monitoring cell density.

$149.00 - $612.00

Specifications

Content And Storage 15°C to 30°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
713003
View Documents
MilliporeSigma™
713003
1 L
Each for $149.00
Only null left
 
713004
View Documents
MilliporeSigma™
713004
5 L
Each for $612.00
Only null left
 
Description

Description

EMD Millipore Novagen™ Overnight Express™ Autoinduction System 1 is based on media components that are metabolized differentially to promote growth.

  • Blend of carbon sources optimized for tightly regulated uninduced growth to high cell density followed by high-level induction
  • Provides magnesium for maximal cell density
  • Convenient for routine expression of proteins in multiple cultures
  • Ideal for high-throughput parallel analysis of protein expression, solubility, purification from multiple expression clones
  • Allows rapid optimization of the host-vector combinations, expression conditions
  • Purification parameters for high-throughput production of proteins for structural or functional analysis
Specifications

Specifications

15°C to 30°C
Videos
SDS
Documents

Documents