Learn More
MilliporeSigma™ Novagen™ NovaQUANT™ Mouse Mitochondrial to Nuclear Ratio kit
Catalog No. 726211KIT
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
726211KIT 1 Kit
1 options
Catalog No. 726211KIT Supplier MilliporeSigma™ Supplier No. 726211KIT
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Compares levels of nuclear to mitochondrial DNA (mtDNA) in mouse DNA sample. It contains set of four optimized PCR primer pairs targeting two nuclear and two mitochondrial genes.
  • qPCR format
  • Can be used whenever small amount of DNA can be extracted from cells or tissues of interest
  • Amplification primers in this kit have been validated for matching amplification efficiency, absence of amplification of mitochondrial pseudogenes
Specifications
Product Type Mouse Mitochondrial to Nuclear Ratio Kit
Content And Storage All materials in this kit can be kept at 4°C for up to three months; Control DNAs at -20°C for long term storage.
Quantity 1 Kit
For Research Use Only