missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ Ni-NTA HisBind™ Resin
Used for rapid one-step purification of proteins.
Supplier: MilliporeSigma™ 706664
Description
EMD Millipore Novagen™ Ni-NTA HisBind™ Resin contains HisTag sequence by metal chelation chromatography.
- High-performance Ni2+-charged agarose
- Minimizes metal leaching during purification
- Binding capacity: 5 to 10mg HisTag™ fusion protein per mL resin
- Supplied as 50% slurry
- Packaging: Glass Bottle
Specifications
| 2°C to 8°C | |
| 25 mL |