missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ KOD XL DNA Polymerase
For PCR amplification of long strand and GC-rich DNA templates
$601.00 - $2,340.00
Specifications
| Concentration | 2.5 U/μL |
|---|---|
| Content And Storage | -20°C |
| Source | Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli (wild type and exonuclease-deficient forms) |
| Product Type | KOD XL DNA Polymerase |
Description
- Used for amplification of large DNA fragments from purified DNA or crude samples
- Amplifies DNA templates up to 30 kbp
- Efficiently incorporates derivatized dNTPs
Specifications
| 2.5 U/μL | |
| -20°C | |
| Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli (wild type and exonuclease-deficient forms) | |
| KOD XL DNA Polymerase |
For Research Use Only