missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ KOD XL DNA Polymerase

Catalog No. 710873
Change view
Click to view available options
Quantity:
1250 U
250 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
710873 250 U
710874 1250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 710873 Supplier MilliporeSigma™ Supplier No. 710873

For PCR amplification of long strand and GC-rich DNA templates

  • Used for amplification of large DNA fragments from purified DNA or crude samples
  • Amplifies DNA templates up to 30 kbp
  • Efficiently incorporates derivatized dNTPs

Specifications

Concentration 2.5 U/μL
Content And Storage -20°C
Source Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli (wild type and exonuclease-deficient forms)
Quantity 250 U
Product Type KOD XL DNA Polymerase

For Research Use Only