missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Novagen™ KOD Hot Start DNA Polymerase
For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications
$462.00 - $1,830.00
Specifications
| Concentration | 1 U/μL |
|---|---|
| Content And Storage | -20°C |
| Source | Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli |
| Product Type | KOD Hot Start DNA Polymerase |
Description
- High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
- Amplifies genomic DNA templates up to 12 kbp
- Amplifies plasmid and lambda DNA templates up to 21 kbp
- Successfully amplifies GC-rich sequences
- Eliminates mispriming and primer-dimer formation
- Convenient room-temperature setup compatible with automation
- Optimal KOD Hot Start Buffer for PCR performance over wide range of targets
Specifications
| 1 U/μL | |
| -20°C | |
| Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli | |
| KOD Hot Start DNA Polymerase |
For Research Use Only