missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ KOD Hot Start DNA Polymerase

Catalog No. 710864
Change view
Click to view available options
Quantity:
1000 U
200 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
710864 1000 U
710863 200 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 710864 Supplier MilliporeSigma™ Supplier No. 710864

For PCR amplification of long strand and GC-rich DNA templates, cloning and cDNA amplification applications

  • High accuracy, yield, and processivity of commercially available proofreading DNA polymerases
  • Amplifies genomic DNA templates up to 12 kbp
  • Amplifies plasmid and lambda DNA templates up to 21 kbp
  • Successfully amplifies GC-rich sequences
  • Eliminates mispriming and primer-dimer formation
  • Convenient room-temperature setup compatible with automation
  • Optimal KOD Hot Start Buffer for PCR performance over wide range of targets

Specifications

Concentration 1 U/μL
Content And Storage -20°C
Source Recombinant Thermococcus kodakaraensis KOD1 DNA polymerase expressed in E. coli
Quantity 1000 U
Product Type KOD Hot Start DNA Polymerase

For Research Use Only