missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Novagen™ BetaRed™ β-Galactosidase Assay Kit

Catalog No. 709783
Change view
Click to view available options
Quantity:
500/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
709783 500/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 709783 Supplier MilliporeSigma™ Supplier No. 709783

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for rapid, sensitive measurement of β-galactosidase activity in cell extracts.

EMD Millipore Novagen™ BetaRed™ β-Galactosidase Assay Kit enables efficient passive extraction of cell protein while maintaining optimal enzyme activity.

  • Configured for 96-well format
  • Colorimetric BetaRed assay is sensitive (1pg)
  • Can be performed in conjunction with luciferase assays

Specifications

For Use With (Application) Measurement of beta-galactosidase activity in cell extracts
Quantity 500/Pk.
Storage Requirements Store at -20°C.