missing translation for 'onlineSavingsMsg'
Learn More

NOVA1 Antibody, ProSci

Numéro de catalogue. 16825420
Modifier l'affichage
Click to view available options
Quantité:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
16825420 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 16825420 Fournisseur ProSci Llc Code fournisseur 466940100

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Goat Polyclonal Antibody

NOVA1 Polyclonal Antibody specifically detects NOVA1 in Human samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Spécifications

Antigène NOVA1
Applications ELISA, Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 500 ug/mL
Conjugué Unconjugated
Dilution Peptide ELISA: antibody detection limit dilution 1:64000.Western Blot:Approx 50-55kDa band observed in Human Breast Cancer lysates (calculated MW of 51.7kDa according to NP_002506.2). In transfected HEK293 transiently expressing NOVA1 a band of approx. 52kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.01-0.03ug/mL. Primary incubation was 1 hour.Immunohistochemistry:Paraffin embedded Human Brain (Cortex) and Heart. Recommended concentration: 5ug/mL.
Formule Supplied at 0.5 mg/mL in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20°C. Minimize freezing and thawing.
Numéro d’acquisition du gène NP_002506.2,NP_006480.2,NP_006482.1
Alias du gène NOVA1, neuro-oncological ventral antigen 1, Nova-1, paraneoplastic Ri antigen, ventral neuron-specific protein 1
Espèces hôtes Goat
Méthode de purification Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Quantité 0.1 mg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 4857
Spécifications de test This antibody is expected to recognise all three reported isoforms (NP_002506.2 ; NP_006480.2 ; NP_006482.1)
Espèces cibles Human
Contenu et stockage Aliquot and store at -20°C. Minimize freezing and thawing.
Type de produit Antibody
Forme Liquid
Isotype IgG
Catégorie de recherche Cancer, Neuroscience
Afficher plus de résultats Afficher moins de résultats