missing translation for 'onlineSavingsMsg'
Learn More

PeproTech NOV Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Numéro de catalogue. p-7245059
Encompass
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
500P257BT25 25 μg
500P257BT50 50 μg
502718562 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 500P257BT25 Fournisseur PeproTech Code fournisseur 500P257BT25UG
Il en reste null

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MQVAATQRCP PQCPGRCPAT PPTCAPGVRA VLDGCSCCLV CARQRGESCS DLEPCDESSG LYCDRSADPS NQTGICTAVE GDNCVFDGVI YRSGEKFQPS CKFQCTCRDG QIGCVPRCQL DVLLPEPNCP APRKVEVPGE CCEKWICGPD EEDSLGGLTL AAYRPEATLG VEVSDSSVNC IEQTTEWTAC SKSCGMGFST RVTNRNRQCE MLKQTRLCMV RPCEQEPEQP TDKKGKKCLR TKKSLKAIHL QFKNCTSLHT YKPRFCGVCS DGRCCTPHNT KTIQAEFQCS PGQIVKKPVM VIGTCTCHTN CPKNNEAFLQ ELELKTTRGK M. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human NOV. Anti-Human NOV-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hNOV by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human NOV (500-P257) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hNOV. Western Blot: To detect hNOV by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hNOV is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions 500-P257BT-1MG will be provided as 2 x 500 μg

NOV is a member of the CCN family of secreted, cysteine-rich regulatory proteins. The full-length NOV protein contains four structural domains that confer distinct, and sometimes opposing, biological activities. Elevated expression of NOV is associated with certain tumors, including Wilm's tumor and most nephroblastomas. However, in other tumor types and certain cancer cell lines, increased tumorigenicity and proliferation is correlated with decreased NOV expression. Additionally, NOV induces cell adhesion and cell migration by signaling through specific cell-surface integrins, and by binding to heparin sulfate proteoglycans and to fibulin 1C. NOV has also been reported to exert proangiogenic activities.

Spécifications

Antigen NOV
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene CCN3
Gene Accession No. P48745
Gene Alias C130088N23Rik; CCN family member 3; CCN3; Cellular communication network factor 3; IBP-9; IGF-binding protein 9; IGFBP9; IGFBP-9; Insulin-like growth factor-binding protein 9; Nephro blastoma-overexpressed gene protein homolog; nephroblastoma overexpressed; nephroblastoma overexpressed gene; nephroblastoma overexpressed gene protein homolog; nephroblastoma-overexpressed gene protein homolog; NOV; NovH; protein NOV homolog
Gene Symbols CCN3
Host Species Rabbit
Immunogen E.coli-derived, 36.2 kDa Recombinant Human NOV.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4856
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Afficher plus de résultats Afficher moins de résultats