missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Human Serum

Catalog No. R00010A
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00010A 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00010A Supplier Invitrogen™ Supplier No. R00010A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Control

Titer results: Gel double diffusion titer : 1:64. Gel double diffusion titer is defined as the dilution of antiserum sufficient to form a precipitate arc with 5 μg antigen and 5 μL per well.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.

Specifications

Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Form Liquid
Product Type Normal Human Serum
Quantity 1.5 mL
Concentration Conc. Not Determined
Formulation Whole serum with 0.1% sodium azide
For Use With (Application) Western Blot, Immunoelectrophoresis, ELISA, Immunodiffusion
Isotype Ig