Learn More
Abcam Inc Normal Horse Serum (sterile), 25ML
Numéro de catalogue. 502653778
Encompass
Horse serum for blocking in antibody-based applications or use as an antibody diluent (ab7484).
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502653778 Fournisseur Abcam Inc Code fournisseur AB748425ML
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Horse serum for blocking in antibody-based applications or use as an antibody diluent (ab7484).
The product is subject to the following: Abcam Restricted Use Statement