Learn More
Abcam Inc Normal Horse Serum (sterile), 50ML
Numéro de catalogue. 502658154
Encompass
Raised in: Horse Purity: Whole antiserum Sterilization: This product was aseptically filtered through a 0.22 micron filter into clean, pre-sterilized containers.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502658154 Fournisseur Abcam Inc Code fournisseur AB13950150ML
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Raised in: Horse Purity: Whole antiserum Sterilization: This product was aseptically filtered through a 0.22 micron filter into clean, pre-sterilized containers.
The product is subject to the following: Abcam Restricted Use Statement