missing translation for 'onlineSavingsMsg'
Learn More

Norlaudanosine Hydrochloride 98.0+%, TCI America™

Catalog No. N09185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09185G 5 g
N091825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09185G Supplier TCI America Supplier No. N09185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6429-04-5
Molecular Formula C20H26ClNO4
Molecular Weight (g/mol) 379.88
MDL Number MFCD00035267
InChI Key VMPLLPIDRGXFTQ-UHFFFAOYNA-N
Synonym Tetrahydropapaverine Hydrochloride, 1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride
PubChem CID 16667431
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES COC1=C(C=C(C=C1)CC2C3=CC(=C(C=C3CCN2)OC)OC)OC.Cl

Specifications

Melting Point 218°C
Color White
Quantity 5 g
Formula Weight 379.88
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Norlaudanosine Hydrochloride
RTECSNumber NX5070700
TSCA No