missing translation for 'onlineSavingsMsg'
Learn More

Norethindrone (Norethisterone) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012235
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012235 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012235 Supplier Merck Emd Millipore Supplier No. PHR17141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Norethindrone (Norethisterone) is a progestin medication, widely used to treat gynecological irregularities such as premenstrual syndrome, painful periods, abnormal heavy bleeding, irregular periods, and menopausal syndrome.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 68-22-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H26O2
MDL Number MFCD00067596
Synonym Norethindrone; 17 alpha-Ethynyl-17 beta-hydroxy-19-nor-4-androsten-3-one; 17 alpha-Ethynyl-19-nortestosterone; 17-Hydroxy-19-nor-17 alpha-4-pregnen-20-yn-3-one; 19-Nor-17 alpha-ethynyl-4-androsten-17 beta-ol-3-one; 19-Norethindrone; 19-Norethisterone
Type Certified Reference Material
Quantity 1 g
Formula Weight 298.42
Grade Certified Reference Material