missing translation for 'onlineSavingsMsg'
Learn More

Norbornadiene Rhodium(I) Chloride Dimer, TCI America™

Catalog No. N0453100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0453100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0453100MG Supplier TCI America Supplier No. N0453100MG

Chemical Identifiers

CAS 12257-42-0
Molecular Formula C14H16Cl2Rh2
Molecular Weight (g/mol) 460.99
MDL Number MFCD00198060
InChI Key RXDWVIOULPVOEO-UHFFFAOYSA-L
Synonym bicyclo 2.2.1 hepta-2,5-diene-rhodium chloride dimer, bicyclo 2.2.1 hepta-2,5-diene; rhodium 2+ ; dichloride, bis 2,3,5,6-eta-bicyclo 2.2.1 hepta-2,5-diene di-mu-chlorodirhodium
PubChem CID 114600
IUPAC Name bis(λ¹-rhodium(1+)) bis(bicyclo[2.2.1]hepta-2,5-diene) dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].C1C2C=CC1C=C2.C1C2C=CC1C=C2

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 460.99
Physical Form Crystalline Powder
Chemical Name or Material Norbornadiene Rhodium(I) Chloride Dimer