missing translation for 'onlineSavingsMsg'
Learn More

(+)-Nootkatone 97.0+%, TCI America™

Catalog No. N09201G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09201G Supplier TCI America Supplier No. N09201G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4674-50-4
Molecular Formula C15H22O
Molecular Weight (g/mol) 218.34
MDL Number MFCD04974578
InChI Key WTOYNNBCKUYIKC-SLEUVZQESA-N
Synonym unii-zms1vjk5hy, zms1vjk5hy, nootkatone,-, +-nootkatone, crystalline, 2 3h-naphthalenone, 4,4a,5,6,7,8-hexahydro-4,4a-dimethyl-6-1-methylethenyl-, 4s,4ar,6s, unii-3k3okv2a5a component, 4s,6s,4ar-4,4a-dimethyl-6-1-methylvinyl-3,4,5,6,7,8,4a-heptahydronaphthale n-2-one
PubChem CID 7567181
IUPAC Name (4S,4aR,6S)-4,4a-dimethyl-6-prop-1-en-2-yl-3,4,5,6,7,8-hexahydronaphthalen-2-one
SMILES CC1CC(=O)C=C2C1(CC(CC2)C(=C)C)C

Specifications

Melting Point 36°C
Color White-Yellow
Quantity 1 g
Formula Weight 218.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (+)-Nootkatone
EINECSNumber (4)-0595
TSCA Yes
Recommended Storage Refrigerator