missing translation for 'onlineSavingsMsg'
Learn More

Nonylphenol diethoxylate Solution, 5 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111016622
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016622 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016622 Supplier Merck Emd Millipore Supplier No. 3289910ML

Nonylphenol diethoxylate (5 μg/mL in acetone) is grouped under the class of alkylphenol ethoxylate surfactants mainlly utilized for households, industry and agriculture.

Environmental

Specifications

CAS 67-64-1
Flash Point -17°C
Linear Formula C19H32O3
Synonym NP-2 EO technical
Type Analytical Standard
Quantity 10 mL
Formula Weight 308.46
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 5 μg/mL in acetone