missing translation for 'onlineSavingsMsg'
Learn More

Nonyl beta-D-glucopyranoside, 98%

Catalog No. AAJ6600903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6600903 1 g
AAJ6600906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6600903 Supplier Thermo Scientific Chemicals Supplier No. J6600903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69984-73-2 | C15H30O6 | 306.399 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69984-73-2
Molecular Formula C15H30O6
Molecular Weight (g/mol) 306.399
MDL Number MFCD00063300
InChI Key QFAPUKLCALRPLH-UXXRCYHCSA-N
Synonym nonyl beta-d-glucopyranoside, b-nonylglucoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy tetrahydro-2h-pyran-3,4,5-triol, b-d-glucopyranoside, nonyl, nonyl-beta-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy oxane-3,4,5-triol, n-nonyl-beta-d-glucopyranoside, bng, beta-d-glucopyranoside, nonyl, 4gby
PubChem CID 155448
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol
SMILES CCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O

Specifications

Color White
Quantity 1 g
Beilstein 12168
Formula Weight 306.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nonyl beta-D-glucopyranoside
EINECSNumber 615-042-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only